(3-Bromobenzo[b]thiophen-2-yl)(cyclohexyl)methanol ≥95%

Supplier: AOBChem USA

Warning

346663-1G 346663-500MG 346663-5G 346663-25G
77470-000EA 415.09 USD
77470-000 77482-668 77493-094 77502-902
(3-Bromobenzo[b]thiophen-2-yl)(cyclohexyl)methanol ≥95%
(3-Bromobenzo[b]thiophen-2-yl)(cyclohexyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₇BrOS
Storage Temperature: Ambient
CAS Number: 3002442-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR