1-(3-Bromobenzo[b]thiophen-2-yl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

346660-1G 346660-500MG 346660-5G 346660-25G
77469-996EA 415.09 USD
77469-996 77482-664 77493-090 77502-898
1-(3-Bromobenzo[b]thiophen-2-yl)-2-fluoroethanone ≥95%
1-(3-Bromobenzo[b]thiophen-2-yl)-2-fluoroethanone

Precaución: For research use only

Formula: C₁₀H₆BrFOS
MW: 273.121 g/mol
Storage Temperature: Ambient
CAS Number: 3002487-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR