1-(3-Bromobenzo[b]thiophen-2-yl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

346658-1G 346658-500MG 346658-5G 346658-25G
77469-992EA 415.09 USD
77469-992 77482-660 77493-086 77502-894
1-(3-Bromobenzo[b]thiophen-2-yl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(3-Bromobenzo[b]thiophen-2-yl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₁H₄BrF₅OS
MW: 359.11 g/mol
Storage Temperature: Ambient
CAS Number: 3002476-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR