1-(3-Bromobenzo[b]thiophen-2-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

346655-1G 346655-500MG 346655-5G 346655-25G
77469-988EA 415.09 USD
77469-988 77482-656 77493-082 77502-890
1-(3-Bromobenzo[b]thiophen-2-yl)propan-1-one ≥95%
1-(3-Bromobenzo[b]thiophen-2-yl)propan-1-one

Precaución: For research use only

Formula: C₁₁H₉BrOS
Storage Temperature: Ambient
CAS Number: 3002478-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR