3-(3-Chloro-2,6-difluorophenyl)pentan-3-ol ≥95%

Supplier: AOBChem USA

Warning

346507-1G 346507-500MG 346507-5G 346507-25G
77469-984EA 415.09 USD
77469-984 77482-652 77493-078 77502-886
3-(3-Chloro-2,6-difluorophenyl)pentan-3-ol ≥95%
3-(3-Chloro-2,6-difluorophenyl)pentan-3-ol

Precaución: For research use only

Formula: C₁₁H₁₃ClF₂O
Storage Temperature: Ambient
CAS Number: 3002486-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR