1-(3-Chloro-2,6-difluorophenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

346505-1G 346505-500MG 346505-5G 346505-25G
77469-980EA 415.09 USD
77469-980 77482-648 77493-074 77502-882
1-(3-Chloro-2,6-difluorophenyl)-2-methylpropan-1-ol ≥95%
1-(3-Chloro-2,6-difluorophenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₁ClF₂O
Storage Temperature: Ambient
CAS Number: 3002502-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR