1-(3-Chloro-2,6-difluorophenyl)-2-ethylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

346501-1G 346501-500MG 346501-5G 346501-25G
77469-976EA 415.09 USD
77469-976 77482-644 77493-070 77502-878
1-(3-Chloro-2,6-difluorophenyl)-2-ethylbutan-1-ol ≥95%
1-(3-Chloro-2,6-difluorophenyl)-2-ethylbutan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₅ClF₂O
MW: 248.697 g/mol
Storage Temperature: Ambient
CAS Number: 3002529-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR