(6-Chloro-2-fluoro-3-methylphenyl)(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

346499-1G 346499-500MG 346499-5G 346499-25G
77469-972EA 415.09 USD
77469-972 77482-640 77493-066 77502-874
(6-Chloro-2-fluoro-3-methylphenyl)(m-tolyl)methanol ≥95%
(6-Chloro-2-fluoro-3-methylphenyl)(m-tolyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₄ClFO
MW: 264.722 g/mol
Storage Temperature: Ambient
CAS Number: 3002441-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR