1-(6-Chloro-2-fluoro-3-methylphenyl)-2-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

346496-1G 346496-500MG 346496-5G 346496-25G
77469-968EA 415.09 USD
77469-968 77482-636 77493-062 77502-870
1-(6-Chloro-2-fluoro-3-methylphenyl)-2-methylbutan-1-ol ≥95%
1-(6-Chloro-2-fluoro-3-methylphenyl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆ClFO
Storage Temperature: Ambient
CAS Number: 3002452-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR