(6-Chloro-2-fluoro-3-methylphenyl)(4-ethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

346492-1G 346492-500MG 346492-5G 346492-25G
77469-964EA 415.09 USD
77469-964 77482-632 77493-058 77502-866
(6-Chloro-2-fluoro-3-methylphenyl)(4-ethylphenyl)methanol ≥95%
(6-Chloro-2-fluoro-3-methylphenyl)(4-ethylphenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₆ClFO
MW: 278.749 g/mol
Storage Temperature: Ambient
CAS Number: 3002506-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR