(6-Chloro-2-fluoro-3-methylphenyl)(3-Chlorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

346488-1G 346488-500MG 346488-5G 346488-25G
77469-956EA 415.09 USD
77469-956 77482-624 77493-050 77502-858
(6-Chloro-2-fluoro-3-methylphenyl)(3-Chlorophenyl)methanol ≥95%
(6-Chloro-2-fluoro-3-methylphenyl)(3-Chlorophenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₁Cl₂FO
Storage Temperature: Ambient
CAS Number: 3002543-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR