(6-Chloro-2-fluoro-3-methylphenyl)(cyclohexyl)methanone ≥95%

Supplier: AOBChem USA

Warning

346483-1G 346483-500MG 346483-5G 346483-25G
77469-952EA 415.09 USD
77469-952 77482-620 77493-046 77502-854
(6-Chloro-2-fluoro-3-methylphenyl)(cyclohexyl)methanone ≥95%
(6-Chloro-2-fluoro-3-methylphenyl)(cyclohexyl)methanone

Precaución: For research use only

Formula: C₁₄H₁₆ClFO
Storage Temperature: Ambient
CAS Number: 3002486-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR