(6-Chloro-2-fluoro-3-methylphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

346480-1G 346480-500MG 346480-5G 346480-25G
77469-948EA 415.09 USD
77469-948 77482-616 77493-042 77502-850
(6-Chloro-2-fluoro-3-methylphenyl)(cyclopropyl)methanol ≥95%
(6-Chloro-2-fluoro-3-methylphenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₁H₁₂ClFO
Storage Temperature: Ambient
CAS Number: 3002452-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR