1-(6-Chloro-2-fluoro-3-methylphenyl)-2-ethylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

346478-1G 346478-500MG 346478-5G 346478-25G
77469-944EA 415.09 USD
77469-944 77482-612 77493-038 77502-846
1-(6-Chloro-2-fluoro-3-methylphenyl)-2-ethylbutan-1-ol ≥95%
1-(6-Chloro-2-fluoro-3-methylphenyl)-2-ethylbutan-1-ol

Precaución: For research use only

Formula: C₁₃H₁₈ClFO
MW: 244.733 g/mol
Storage Temperature: Ambient
CAS Number: 3002529-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR