(6-Chloro-2-fluoro-3-methylphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

346476-1G 346476-500MG 346476-5G 346476-25G
77469-940EA 415.09 USD
77469-940 77482-608 77493-034 77502-842
(6-Chloro-2-fluoro-3-methylphenyl)triethylsilane ≥95%
(6-Chloro-2-fluoro-3-methylphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₃H₂₀ClFSi
MW: 258.835 g/mol
Storage Temperature: Ambient
CAS Number: 3002543-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR