1-Butyl-4-ethylbenzene ≥95%

Supplier: AOBChem USA

Warning

346364-1G 346364-500MG 346364-5G 346364-25G
77469-936EA 415.09 USD
77469-936 77482-604 77493-030 77502-838
1-Butyl-4-ethylbenzene ≥95%
1-Butyl-4-ethylbenzene

Precaución: For research use only

Formula: C₁₂H₁₈
Storage Temperature: Ambient
CAS Number: 15181-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR