1-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)-2-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

346276-1G 346276-500MG 346276-5G 346276-25G
77469-932EA 415.09 USD
77469-932 77482-600 77493-026 77502-834
1-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)-2-methylbutan-1-ol ≥95%
1-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)-2-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₇H₁₈BrFO
MW: 337.2266 g/mol
Storage Temperature: Ambient
CAS Number: 3002452-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR