1-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

346273-1G 346273-500MG 346273-5G 346273-25G
77469-928EA 415.09 USD
77469-928 77482-596 77493-022 77502-830
1-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)prop-2-en-1-ol ≥95%
1-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)prop-2-en-1-ol

Precaución: For research use only

Formula: C₁₅H₁₂BrFO
MW: 307.1575 g/mol
Storage Temperature: Ambient
CAS Number: 3002504-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR