2-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)pentan-2-ol ≥95%

Supplier: AOBChem USA

Warning

346271-1G 346271-500MG 346271-5G 346271-25G
77469-924EA 415.09 USD
77469-924 77482-592 77493-018 77502-826
2-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)pentan-2-ol ≥95%
2-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)pentan-2-ol

Precaución: For research use only

Formula: C₁₇H₁₈BrFO
MW: 337.2266 g/mol
Storage Temperature: Ambient
CAS Number: 3002543-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR