1-([2,2'-bithiophen]-5-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

346268-1G 346268-500MG 346268-5G 346268-25G
77469-920EA 415.09 USD
77469-920 77482-588 77493-014 77502-822
1-([2,2'-bithiophen]-5-yl)-2,2-difluoroethanone ≥95%
1-([2,2'-bithiophen]-5-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₆F₂OS₂
Storage Temperature: Ambient
CAS Number: 3002441-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR