1-Fluoro-2-methoxy-4-(methylsulfanyl)benzene ≥97%

Supplier: AOBChem USA

Warning

34600-1G 34600-500MG 34600-5G 34600-25G
77469-912EA 146.26 USD
77469-912 77482-580 77493-006 77502-816
1-Fluoro-2-methoxy-4-(methylsulfanyl)benzene ≥97%
1-Fluoro-2-methoxy-4-(methylsulfanyl)benzene

Precaución: For research use only

Formula: C₈H₉FOS
Storage Temperature: Ambient
MDL Number: MFCD22489311
CAS Number: 1822763-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR