4-(methoxymethoxy)-2,3-dimethylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

34536-1G 34536-500MG 34536-5G 34536-25G
77469-904EA 282.22 USD
77469-904 77482-572 77492-998 77502-812
4-(methoxymethoxy)-2,3-dimethylbenzaldehyde ≥95%
4-(methoxymethoxy)-2,3-dimethylbenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902587
CAS Number: 2586127-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR