(3,5-Di-tert-butylphenyl)(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

345353-1G 345353-500MG 345353-5G 345353-25G
77469-900EA 415.09 USD
77469-900 77482-568 77492-994 77502-808
(3,5-Di-tert-butylphenyl)(m-tolyl)methanol ≥95%
(3,5-Di-tert-butylphenyl)(m-tolyl)methanol

Precaución: For research use only

Formula: C₂₂H₃₀O
Storage Temperature: Ambient
CAS Number: 3002460-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR