3,4-Dichloro-2-iodoanisole ≥95%

Supplier: AOBChem USA

Warning

34446-1G 34446-500MG 34446-5G 34446-25G
77469-896EA 250.29 USD
77469-896 77482-564 77492-986 77502-800
3,4-Dichloro-2-iodoanisole ≥95%
3,4-Dichloro-2-iodoanisole

Precaución: For research use only

Formula: C₇H₅Cl₂IO
MW: 302.924 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619491
CAS Number: 873386-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR