Ethyl 2,3-Dichloro-6-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

34373-1G 34373-500MG 34373-5G
77469-892EA 435.69 USD
77469-892 77482-560 77492-982
Ethyl 2,3-Dichloro-6-methoxybenzoate ≥97%
Ethyl 2,3-Dichloro-6-methoxybenzoate

Precaución: For research use only

Formula: C₁₀H₁₀Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD31619484
CAS Number: 2179038-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR