(E)-isopropyl 3-hydroxyhex-4-enoate ≥95%

Supplier: AOBChem USA

Warning

343080-1G 343080-500MG 343080-5G 343080-25G
77469-884EA 415.09 USD
77469-884 77482-552 77492-974 77502-792
(E)-isopropyl 3-hydroxyhex-4-enoate ≥95%
(E)-isopropyl 3-hydroxyhex-4-enoate

Precaución: For research use only

Formula: C₉H₁₆O₃
MW: 172.2215 g/mol
Storage Temperature: Ambient
CAS Number: 41096-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR