Isopropyl 3-hydroxy-3-(3-methoxyphenyl)propanoate ≥95%

Supplier: AOBChem USA

Warning

343077-1G 343077-500MG 343077-5G 343077-25G
77469-880EA 415.09 USD
77469-880 77482-548 77492-970 77502-788
Isopropyl 3-hydroxy-3-(3-methoxyphenyl)propanoate ≥95%
Isopropyl 3-hydroxy-3-(3-methoxyphenyl)propanoate

Precaución: For research use only

Formula: C₁₃H₁₈O₄
MW: 238.2796 g/mol
Storage Temperature: Ambient
CAS Number: 3002486-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR