Isopropyl 3-(3-chlorophenyl)-3-hydroxypropanoate ≥95%

Supplier: AOBChem USA

Warning

343072-1G 343072-500MG 343072-5G 343072-25G
77469-872EA 415.09 USD
77469-872 77482-540 77492-962 77502-780
Isopropyl 3-(3-chlorophenyl)-3-hydroxypropanoate ≥95%
Isopropyl 3-(3-chlorophenyl)-3-hydroxypropanoate

Precaución: For research use only

Formula: C₁₂H₁₅ClO₃
Storage Temperature: Ambient
CAS Number: 3002441-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR