Isopropyl 2-(1-hydroxycyclopentyl)acetate ≥95%

Supplier: AOBChem USA

Warning

343068-1G 343068-500MG 343068-5G 343068-25G
77469-864EA 347.11 USD
77469-864 77482-532 77492-954 77502-772
Isopropyl 2-(1-hydroxycyclopentyl)acetate ≥95%
Isopropyl 2-(1-hydroxycyclopentyl)acetate

Precaución: For research use only

Formula: C₁₀H₁₈O₃
Storage Temperature: Ambient
CAS Number: 1866283-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR