Isopropyl 2-(1-hydroxycyclobutyl)acetate ≥95%

Supplier: AOBChem USA

Warning

343066-1G 343066-500MG 343066-5G 343066-25G
77469-860EA 415.09 USD
77469-860 77482-528 77492-950 77502-768
Isopropyl 2-(1-hydroxycyclobutyl)acetate ≥95%
Isopropyl 2-(1-hydroxycyclobutyl)acetate

Precaución: For research use only

Formula: C₉H₁₆O₃
Storage Temperature: Ambient
CAS Number: 3002476-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR