(3-(tert-Butyl)phenyl)(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

343063-1G 343063-500MG 343063-5G 343063-25G
77469-856EA 415.09 USD
77469-856 77482-524 77492-946 77502-766
(3-(tert-Butyl)phenyl)(m-tolyl)methanol ≥95%
(3-(tert-Butyl)phenyl)(m-tolyl)methanol

Precaución: For research use only

Formula: C₁₈H₂₂O
MW: 254.3666 g/mol
Storage Temperature: Ambient
CAS Number: 3002460-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR