(3-(tert-Butyl)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

343061-1G 343061-500MG 343061-5G
77469-852EA 668.47 USD
77469-852 77482-520 77492-942
(3-(tert-Butyl)phenyl)(methyl)sulfane ≥95%
(3-(tert-Butyl)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₆S
Storage Temperature: Ambient
CAS Number: 1314966-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR