(2,3-Difluorophenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

343040-1G 343040-500MG 343040-5G 343040-25G
77469-848EA 415.09 USD
77469-848 77482-516 77492-938 77502-762
(2,3-Difluorophenyl)triethylsilane ≥95%
(2,3-Difluorophenyl)triethylsilane

Precaución: For research use only

Formula: C₁₂H₁₈F₂Si
MW: 228.3536 g/mol
Storage Temperature: Ambient
CAS Number: 3002501-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR