(2-Bromo-6-fluoro-4-methylphenyl)(2-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

342666-1G 342666-500MG 342666-5G 342666-25G
77469-844EA 415.09 USD
77469-844 77482-512 77492-934 77502-758
(2-Bromo-6-fluoro-4-methylphenyl)(2-methoxyphenyl)methanol ≥95%
(2-Bromo-6-fluoro-4-methylphenyl)(2-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₄BrFO₂
MW: 325.1728 g/mol
Storage Temperature: Ambient
CAS Number: 3002520-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR