(2-Bromo-6-fluoro-4-methylphenyl)(3,5-dimethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

342663-1G 342663-500MG 342663-5G 342663-25G
77469-840EA 415.09 USD
77469-840 77482-508 77492-930 77502-754
(2-Bromo-6-fluoro-4-methylphenyl)(3,5-dimethylphenyl)methanol ≥95%
(2-Bromo-6-fluoro-4-methylphenyl)(3,5-dimethylphenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₆BrFO
MW: 323.2000 g/mol
Storage Temperature: Ambient
CAS Number: 3002543-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR