(2-Bromo-6-fluoro-4-methylphenyl)(4-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

342661-1G 342661-500MG 342661-5G 342661-25G
77469-836EA 415.09 USD
77469-836 77482-504 77492-926 77502-750
(2-Bromo-6-fluoro-4-methylphenyl)(4-fluorophenyl)methanol ≥95%
(2-Bromo-6-fluoro-4-methylphenyl)(4-fluorophenyl)methanol

Precaución: For research use only

Formula: C₁₄H₁₁BrF₂O
MW: 313.1373 g/mol
Storage Temperature: Ambient
CAS Number: 3002483-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR