3-(2-Bromo-6-fluoro-4-methylphenyl)pentan-3-ol ≥95%

Supplier: AOBChem USA

Warning

342658-1G 342658-500MG 342658-5G 342658-25G
77469-832EA 415.09 USD
77469-832 77482-500 77492-922 77502-746
3-(2-Bromo-6-fluoro-4-methylphenyl)pentan-3-ol ≥95%
3-(2-Bromo-6-fluoro-4-methylphenyl)pentan-3-ol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO
MW: 275.1572 g/mol
Storage Temperature: Ambient
CAS Number: 3002520-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR