(2-Bromo-6-fluoro-4-methylphenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

342644-1G 342644-500MG 342644-5G 342644-25G
77469-824EA 415.09 USD
77469-824 77482-490 77492-912 77502-738
(2-Bromo-6-fluoro-4-methylphenyl)triethylsilane ≥95%
(2-Bromo-6-fluoro-4-methylphenyl)triethylsilane

Precaución: For research use only

Formula: C₁₃H₂₀BrFSi
Storage Temperature: Ambient
CAS Number: 3002441-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR