(4-(tert-Butyl)phenyl)(cyclohexyl)methanone ≥95%

Supplier: AOBChem USA

Warning

342601-1G 342601-500MG 342601-5G
77469-820EA 594.31 USD
77469-820 77482-488 77492-910
(4-(tert-Butyl)phenyl)(cyclohexyl)methanone ≥95%
(4-(tert-Butyl)phenyl)(cyclohexyl)methanone

Precaución: For research use only

Formula: C₁₇H₂₄O
Storage Temperature: Ambient
CAS Number: 28047-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR