2-Chloro-1-(4'-ethyl-[1,1'-biphenyl]-4-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

342223-1G 342223-500MG 342223-5G 342223-25G
77469-816EA 415.09 USD
77469-816 77482-484 77492-906 77502-734
2-Chloro-1-(4'-ethyl-[1,1'-biphenyl]-4-yl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(4'-ethyl-[1,1'-biphenyl]-4-yl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₆H₁₃ClF₂O
MW: 294.724 g/mol
Storage Temperature: Ambient
CAS Number: 3002543-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR