(4'-ethyl-[1,1'-biphenyl]-4-yl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

342219-1G 342219-500MG 342219-5G 342219-25G
77469-812EA 415.09 USD
77469-812 77482-480 77492-902 77502-730
(4'-ethyl-[1,1'-biphenyl]-4-yl)trimethylsilane ≥95%
(4'-ethyl-[1,1'-biphenyl]-4-yl)trimethylsilane

Precaución: For research use only

Formula: C₁₇H₂₂Si
Storage Temperature: Ambient
CAS Number: 2850226-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR