1-(2,6-Dichloro-5-methylpyridin-3-yl)-2-ethylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

342188-1G 342188-500MG 342188-5G 342188-25G
77469-808EA 415.09 USD
77469-808 77482-476 77492-898 77502-726
1-(2,6-Dichloro-5-methylpyridin-3-yl)-2-ethylbutan-1-ol ≥95%
1-(2,6-Dichloro-5-methylpyridin-3-yl)-2-ethylbutan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₇Cl₂NO
MW: 262.175 g/mol
Storage Temperature: Ambient
CAS Number: 3002441-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR