(4-(tert-Butyl)phenyl)(3-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

341952-1G 341952-500MG 341952-5G 341952-25G
77469-804EA 415.09 USD
77469-804 77482-472 77492-894 77502-722
(4-(tert-Butyl)phenyl)(3-methoxyphenyl)methanol ≥95%
(4-(tert-Butyl)phenyl)(3-methoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₈H₂₂O₂
MW: 270.3660 g/mol
Storage Temperature: Ambient
CAS Number: 883809-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR