Butyl(2-fluoro-5-methylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

341948-1G 341948-500MG 341948-5G 341948-25G
77469-800EA 415.09 USD
77469-800 77482-468 77492-890 77502-718
Butyl(2-fluoro-5-methylphenyl)sulfane ≥95%
Butyl(2-fluoro-5-methylphenyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₅FS
MW: 198.3002 g/mol
Storage Temperature: Ambient
CAS Number: 3002482-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR