1-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)-2-ethylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

341907-1G 341907-500MG 341907-5G 341907-25G
77469-796EA 415.09 USD
77469-796 77482-464 77492-886 77502-714
1-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)-2-ethylbutan-1-ol ≥95%
1-(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)-2-ethylbutan-1-ol

Precaución: For research use only

Formula: C₁₈H₂₀BrFO
MW: 351.253 g/mol
Storage Temperature: Ambient
CAS Number: 3002488-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR