(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)triethylsilane ≥95%

Supplier: AOBChem USA

341901-1G 341901-500MG 341901-5G 341901-25G
77469-792EA 415.09 USD
77469-792 77482-460 77492-882 77502-710
(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)triethylsilane ≥95%
(4-Bromo-2-fluoro-[1,1'-biphenyl]-3-yl)triethylsilane

Precaución: For research use only

Formula: C₁₈H₂₂BrFSi
MW: 365.355 g/mol
CAS Number: 3002439-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR