(E)-1-(perfluorophenyl)but-2-en-1-ol ≥95%

Supplier: AOBChem USA

Warning

341877-1G 341877-500MG 341877-5G 341877-25G
77469-788EA 415.09 USD
77469-788 77482-456 77492-878 77502-708
(E)-1-(perfluorophenyl)but-2-en-1-ol ≥95%
(E)-1-(perfluorophenyl)but-2-en-1-ol

Precaución: For research use only

Formula: C₁₀H₇F₅O
MW: 238.1539 g/mol
Storage Temperature: Ambient
CAS Number: 73727-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR