(perfluorophenyl)(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

Warning

341870-1G 341870-500MG 341870-5G 341870-25G
77469-776EA 415.09 USD
77469-776 77482-444 77492-866 77502-698
(perfluorophenyl)(m-tolyl)methanol ≥95%
(perfluorophenyl)(m-tolyl)methanol

Precaución: For research use only

Formula: C₁₄H₉F₅O
MW: 288.2126 g/mol
Storage Temperature: Ambient
CAS Number: 1226497-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR