1-Methyl-4-(methylthio)-1H-pyrazole ≥95%

Supplier: AOBChem USA

Warning

341854-1G 341854-500MG 341854-5G
77469-772EA 633.45 USD
77469-772 77482-440 77492-862
1-Methyl-4-(methylthio)-1H-pyrazole ≥95%
1-Methyl-4-(methylthio)-1H-pyrazole

Precaución: For research use only

Formula: C₅H₈N₂S
MW: 128.1954 g/mol
Storage Temperature: Ambient
CAS Number: 1309388-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR