1-Bromo-4-(methoxymethoxy)-2-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

34168-1G 34168-500MG 34168-5G 34168-25G
77469-768EA 73.13 USD
77469-768 77482-436 77492-858 77502-694
1-Bromo-4-(methoxymethoxy)-2-(trifluoromethoxy)benzene ≥95%
1-Bromo-4-(methoxymethoxy)-2-(trifluoromethoxy)benzene

Precaución: For research use only

Formula: C₉H₈BrF₃O₃
MW: 301.057 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619468
CAS Number: 2158298-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR